Learn More
Novus Biologicals™ Rat TNF-alpha ELISA Kit [Biotin]
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP192681
Catalog No. NBP192681
Add to cart
Description
Description
The Rat TNF-alpha ELISA Kit (Colorimetric) measures rat TNF-alpha in cell culture supernatant, serum.
Specifications
Specifications
39.1 - 2,500pg/mL
7124
ELISA Kit
11.0pg/mL
Rat (Rt)
TNF-alpha
TNF
ELISA
Aluminium pouch(es) with a Microwell Plate coated with polyclonal antibody to rat TNF-alpha, Biotin-Conjugate anti-rat TNF-alpha polyclonal antibody, Streptavidin-HRP, Rat TNF-alpha Standard lyophilized, 5ng/mL upon reconstitution, Sample Diluent, Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA), Wash Buffer Concentrate 20x (PBS with 1% Tween 20), Substrate Solution (tetramethyl-benzidine), Stop Solution (1M Phosphoric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum
Storage of components varies. See protocol for specific instructions.
Adaptive Immunity, Asthma, Autophagy, Biologically Active Proteins, Cancer, Cytokine Research, Diabetes Research, Immune System Diseases, Immunology, Innate Immunity
APC1 protein, Cachectin, DIF, TNF, monocyte-derived, TNF-a, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necr