Learn More
Crystal Chem IncSupplier Diversity Partner Rat Total Bile Acids Assay Kit, 96-tests DFS Item
Manufacturer:  Crystal Chem IncSupplier Diversity Partner 80460
The Rat Total Bile Acids Assay is used to quantify levels of total bile acids in rat serum and plasma in less than 10 min. Highlights: Kit uses sample size of only 20 µL. Excellent correlation to ELISA method. Fast, 10 minute procedure. Protocol for liver and feces samples available. Designed for rat samples.Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7710