Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Rat TRANCE/RANK L/TNFSF11 Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

$343.50

Specifications

Product Type Recombinant protein
Molecular Weight 29 kDa
Purity >85%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Conjugate Unlabeled
Recombinant Yes
View More Specs

Products 2
Catalog Number Mfr. No. Formulation Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Price Quantity & Availability  
93-66T-N025 View Documents R&D Systems™
9366TN025
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and EDTA with BSA as a carrier protein. Each for $343.50
Add to cart
 
93-66T-N025CF View Documents R&D Systems™
9366TN025/CF
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and EDTA. Each for $343.50
Add to cart
 
Description

Description

The Recombinant Rat TRANCE/RANK L/TNFSF11 Protein is derived from CHO. The Recombinant Rat TRANCE/RANK L/TNFSF11 Protein has been validated for the following applications: Bioactivity.

Order Info

Ships at ambient temperature.

Specifications

Specifications

Recombinant protein
>85%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Yes
<0.10 EU per 1μg of the protein by the LAL method.
XP_008769150
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
29 kDa
Unlabeled
Rat
2.5 to 15ng/mL
RUO
Chinese Hamster Ovary cell line, CHO-derived Arg72-Asp318, with an N-terminal 6-His tag

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit