Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rat Triiodothyronine ELISA Kit (Colorimetric)

Assay has high sensitivity and specificity for detection of rat T3

$501.50

Specifications

Species Rat
Sensitivity 0.5ng/mL
Sample Type serum, plasma, cell culture supernates, tissue homogenates
Research Discipline Signal Transduction
Specificity For detection of rat T3
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601851KT View Documents Novus Biologicals™
NBP2601851KIT
1 Kit Each for $501.50
Add to cart
 
Description

Description

The Rat Triiodothyronine ELISA Kit (Colorimetric) measures rat Triiodothyronine in serum, plasma, cell culture supernates, tissue homogenates.
Specifications

Specifications

Rat
serum, plasma, cell culture supernates, tissue homogenates
For detection of rat T3
Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
ELISA
0.5ng/mL to 8ng/mL
0.5ng/mL
Signal Transduction
liothyronine; T3
Triiodothyronine
ELISA Kit (Colorimetric)

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit