Learn More
Rat TRPV4, Synthetic Peptide, Invitrogen™
Synthetic Peptide
Manufacturer:  Invitrogen GST00012P
Catalog No. PIGST00012P
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Rat TRPV4 Synthetic Peptide, GST00012P, from Invitrogen.
GST00012P is the immunizing peptide for the rabbit antibody to rat TRPV4 (OSR00062W).GST00012P may be used as a control in various immunology applications.Reconstitute with PBS or distilled water.
Specifications
Specifications
Rat
Synthetic Peptide
Rat
Control
peptide with no preservative
RUO
No preservative
Rat TRPV4
Q9ERZ8
Peptide
66026
purified
50μg
Lyophilized
Y
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Transient receptor potential cation channel subfamily V member 4, TrpV4, osm-9-like TRP channel 4, OTRPC4, Vanilloid receptor-related, smotically-acti
Trpv4
Control