Learn More
BioworldSupplier Diversity Partner RAT VASCULAR ENDOTHELIAL GR DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220607471
RAT VASCULAR ENDOTHELIAL GROWTH FACTOR-C (152), (RECOMBINANT): Recombinant Rat Vascular Endothelial Growth Factor-C 152 a.a. (VEGF-C152) Source: Sf9, Insect Cells Background: VEGF-C152S is a point mutant generated by the replacement ...
Catalog No. 50-253-848