Learn More
Raybiotech IncSupplier Diversity Partner Rat VEGF-A ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELRVEGFCL1
For cell and tissue lysate samples. Sensitivity: 2 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes rat VEGF.
Catalog No. 50-195-0315