Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Rat Vitamin A ELISA Kit (Colorimetric)
Assay has high sensitivity and specificity for detection of rat VA
Supplier: Novus Biologicals™ NBP2601941KIT
Description
The Rat Vitamin A ELISA Kit (Colorimetric) measures rat Vitamin A in serum, plasma.Specifications
312.5 pg/ml to 5000 pg/ml | |
ELISA Kit (Colorimetric) | |
For detection of rat VA | |
ELISA | |
1 Kit | |
50 to 100 μL | |
Vitamin A |
233.598pg/mL | |
Plasma, Serum | |
Rat | |
Instruction manual, Stop Solution, Sample Diluent, Standard (10 x concentrate), Wash Buffer (25 x concentrate), TMB Substrate, HRP-conjugate Diluent, HRP-conjugate (100 x concentrate), Adhesive Strip (For 96 wells), Assay plate | |
Lipid and Metabolism | |
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction