Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rat Vitamin A ELISA Kit (Colorimetric)

Assay has high sensitivity and specificity for detection of rat VA

$732.50

Specifications

Sensitivity 233.598pg/mL
Sample Type Plasma, Serum
Research Discipline Lipid and Metabolism
Specificity For detection of rat VA
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601941KT View Documents Novus Biologicals™
NBP2601941KIT
1 Kit Each for $732.50
Add to cart
 
Description

Description

The Rat Vitamin A ELISA Kit (Colorimetric) measures rat Vitamin A in serum, plasma.
Specifications

Specifications

233.598pg/mL
Lipid and Metabolism
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
ELISA
312.5 pg/ml to 5000 pg/ml
Instruction manual, Stop Solution, Sample Diluent, Standard (10 x concentrate), Wash Buffer (25 x concentrate), TMB Substrate, HRP-conjugate Diluent, HRP-conjugate (100 x concentrate), Adhesive Strip (For 96 wells), Assay plate
Plasma, Serum
For detection of rat VA
Vitamin A
ELISA Kit (Colorimetric)
50 to 100 μL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit