Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rats, Fisher Science Education™ Fisher-Free™
SCI_ED

Rattus norvegicus.

Supplier:  Ranaco Corporation 140.1179.1

 View more versions of this product

Catalog No. S1179S


Add to Cart

Description

Description

Rats are similar in price to fetal pigs, but have adult musculature. If you desire a mammal specimen that is inexpensive, fully muscled, and has well-developed genitalia, the rat is the specimen of choice. Pregnant rats are also available.

Order Info

INJECTION KEY
Plain: Circulatory system not injected with colors
Single: Arteries injected with red latex
Double: Arteries red, veins blue
Triple: Arteries red, veins blue, and hepatic portal veins yellow

Specifications

Specifications

Rat
Single
17.78 to 22.86 cm
Large
Fisher-Free™
7 to 9 in.
Ea.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit