Learn More
Restek™ RAVEqc Quick-Connect Air Valves
Simplify workflow with this tool-free design that makes it fast and easy for anyone to make consistent, quality connections
$58.50 - $158.00
Catalog Number | Mfr. No. | Type | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Material | Price | Quantity & Availability | ||||
06-611-872
|
Restek™
27370 |
Female RAVEqc Valve to 1/4 in. Male Compression Fitting | Siltek™ Treated |
Each for $158.00
|
|
||||
06-611-873
|
Restek™
27369 |
Female RAVEqc Valve to 1/4 in. Male Compression Fitting | Stainless Steel |
Each for $147.00
|
|
||||
06-611-886
|
Restek™
27366 |
Female RAVEqc Valve to 1/4 in. NPT Fitting | Siltek™ Treated |
Each for $158.00
|
|
||||
06-611-885
|
Restek™
27365 |
Female RAVEqc Valve to 1/4 in. NPT Fitting | Stainless Steel |
Each for $147.00
|
|
||||
06-611-874
|
Restek™
27368 |
Male RAVEqc Valve to 1/4 in. Male Compression Fitting | Siltek™ Treated |
Each for $113.00
|
|
||||
06-611-875
|
Restek™
27367 |
Male RAVEqc Valve to 1/4 in. Male Compression Fitting | Stainless Steel |
Each for $88.00
|
|
||||
06-611-868
|
Restek™
27374 |
Male RAVEqc Valve to 1/4 in. Tube End Fitting, Short | Siltek™ Treated |
Each for $82.00
|
|
||||
06-611-869
|
Restek™
27373 |
Male RAVEqc Valve to 1/4 in. Tube End Fitting, Short | Stainless Steel |
Each for $58.50
|
|
||||
06-611-870
|
Restek™
27372 |
Male RAVEqc Valve to 1/4 in. Tube End Fitting, Extended | Siltek™ Treated |
Each for $82.00
|
|
||||
06-611-871
|
Restek™
27371 |
Male RAVEqc Valve to 1/4 in. Tube End Fitting, Extended | Stainless Steel |
Each for $68.50
|
|
||||
Description
- Extend Air Canister and Valve Lifetime: No wrenches mean no overtightening or cross-threading damage
- Standalone or Supporting Roles: Use alone or pair with a bellows/diaphragm valve for the ultimate in sample security and convenience
- Check Status at a Glance: If the parts are connected, the valve is open; if they’re apart, it’s closed
- Leak-Free Performance: Each and every valve is helium leak tested to 1x10-6 mL/sec
- Standard compression or tube end fittings accommodate a wide variety of air canisters and related devices
- Compatible with Entech Micro-QT Valves
- A RAVEqc Valve Bracket is required for use with Restek 1, 3, 6, and 15 L Air Sampling Canisters when using the compression fitting mounted directly on the 1/4 in. canister tube stub
Specifications
Flow: 2,500 mL/min @ 29 in. Hg | |
1 |