Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAX, Rabbit anti-Human,Canine,Primate, DyLight 650, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB10056467C

Catalog No. NB003564


Add to cart

Description

Description

RAX Polyclonal specifically detects RAX in Human, Canine, Primate samples. It is validated for Western Blot.
Specifications

Specifications

RAX
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
MCOP3, retina and anterior neural fold homeobox, retinal homeobox protein Rx, RXRetina and anterior neural fold homeobox protein
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human, Canine, Primate
Western Blot
Western Blot
Affinity Purified, Conjugated
RAX
A portion of amino acids 300-350 of human Rax was used as the immunogen.
Immunogen affinity purified
RUO
Primary
The amino acid sequence used as immunogen has 100% identity in human, dog, and monkey.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit