Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Razor Blades
SCI_ED

Single-edged surgical carbon-steel blades are easily and safely gripped.

$74.30

Specifications

Length (English) 1.5 in.
Length (Metric) 3.81 cm
Product Type Razor Blade
Blade Material Carbon Steel
View More Specs

Products 2
Catalog Number Mfr. No. Thickness (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Thickness (Metric) Price Quantity & Availability  
S17302
SCI_ED
View Documents
Accutec Blades Inc
9401150000
0.2 mm
Pack of 100 for $74.30
Case of 50 PK for $3,380.00
Add to cart
 
S65921
SCI_ED
View Documents
Accutec Blades Inc
9501250000
0.3 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

  • Dimensions: 1.5L × 0.75 in.W (3.8 × 1.9cm)

Order Info

Individually shelled in packs of 100.

Specifications

Specifications

1.5 in.
3.81 cm
Razor Blade
Carbon Steel
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit