Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB1 Mouse anti-Human, Mouse, Clone: 1F8, Novus Biologicals™

Mouse Monoclonal Antibody

$152.22 - $378.35

Specifications

Antigen RB1
Clone 1F8
Host Species Mouse
Isotype IgG1 Kappa
Gene Alias OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25339820 View Documents Novus Biologicals
NBP25339820UG
20 ug Each for $152.22
Add to cart
 
NB253398100 View Documents Novus Biologicals
NBP253398100UG
100 ug Each for $378.35
Add to cart
 
NB253398200 View Documents Novus Biologicals
NBP253398200UG
200 ug This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

RB1 Monoclonal antibody specifically detects RB1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

RB1
Mouse
OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
Store at 4C.
Monoclonal
0.2 mg/mL
1F8
IgG1 Kappa
Human, Mouse
RUO
Purified
Primary
10mM PBS with 0.05% BSA with 0.05% Sodium Azide

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit