Learn More
RB1 (phospho S811) Rabbit anti-Human, Mouse, Rat, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human RB1.
Supplier: Abnova Corporation PAB29596
Description
The protein encoded by this gene is a negative regulator of the cell cycle and was the first tumor suppressor gene found. The encoded protein also stabilizes constitutive heterochromatin to maintain the overall chromatin structure. The active, hypophosphorylated form of the protein binds transcription factor E2F1. Defects in this gene are a cause of childhood cancer retinoblastoma (RB), bladder cancer, and osteogenic sarcoma. [provided by RefSeq]
Specifications
RB1 (phospho S811) | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of human RB1. | |
In PBS (without Mg2+ and Ca2+), 150 mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
RB1 | |
A synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S811 of human RB1. | |
100 μL | |
Primary | |
RB1 (phospho S811) polyclonal antibody detects endogenous levels of human RB1 only when phosphorylated at serine 811. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunofluorescence, Western Blot | |
Unconjugated | |
Immunofluorescence (1:50-1:200) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
OSRC/RB/p105-Rb/pRb/pp110 | |
Rabbit | |
Affinity chromatography | |
RUO | |
5925 | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.