Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB1 Rat anti-Human, Mouse, DyLight™ 488, Clone: 7F10, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP275951G

Catalog No. NBP275951G


Add to cart

Description

Description

RB1 Monoclonal antibody specifically detects RB1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

RB1
7F10
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Conjugated, Purified
RB1
Synthetic peptide COOH-C-ERERpTPRKNN-NH2 coupled to mycobacterium tuberculosis PPD
Protein G purified
Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors
Primary
5925.0
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein
Rat
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit