Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB1CC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Encompass_Preferred

$395.30

Specifications

Antigen RB1CC1
Concentration 0.28 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-172-5998
Encompass_Preferred
View Documents Promotion Details
Proteintech Group IncSupplier Diversity Partner
100691AP150UL
150 μL
Each for $395.30
Add to cart
 
Description

Description

This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human RB1CC1.

The protein encoded by this gene interacts with signaling pathways to coordinately regulate cell growth, cell proliferation, apoptosis, autophagy, and cell migration. This tumor suppressor also enhances retinoblastoma 1 gene expression in cancer cells. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications

Specifications

RB1CC1
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Polyclonal
-20°C
0.28 mg/mL
Unconjugated
Rabbit
Human, Mouse
IgG
Antigen Affinity Chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit