Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB1CC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Manufacturer: Proteintech Group Inc 100432AP150UL

Description
The immunogen of this antibody is C-terminal 154aa of RB1CC1.
The protein encoded by this gene interacts with signaling pathways to coordinately regulate cell growth, cell proliferation, apoptosis, autophagy, and cell migration. This tumor suppressor also enhances retinoblastoma 1 gene expression in cancer cells. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Specifications
RB1CC1 | |
Polyclonal | |
Unconjugated | |
RB1CC1 | |
RB1CC1 | |
Rabbit | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.62 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8TDY2, Q9ESK9 | |
RB1CC1 | |
Antigen Affinity Chromatography | |
RUO | |
12421, 312927, 9821 | |
Antibody | |
IgG |
Safety and Handling
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title