Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBAK Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540627
Description
Peptide sequence: HYRSHLEEKP YECNECGKTF NLNSAFIRHR KVHTEEKSHE CSECGKFSQL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 79%; Rat: 100%.
RBAK encodes a nuclear protein which interacts with the tumor suppressor retinoblastoma 1. The two interacting proteins are thought to act as a transcriptional repressor for promoters which are activated by the E2F1 transcription factor. This protein contains a Kruppel-associated box (KRAB), which is a transcriptional repressor motif.This gene encodes a nuclear protein which interacts with the tumor suppressor retinoblastoma 1. The two interacting proteins are thought to act as a transcriptional repressor for promoters which are activated by the E2F1 transcription factor. This protein contains a Kruppel-associated box (KRAB), which is a transcriptional repressor motif.
Specifications
RBAK | |
Polyclonal | |
Unconjugated | |
RBAK | |
hRBaK; RB associated KRAB zinc finger; RBAK; RB-associated KRAB repressor; RB-associated KRAB zinc finger; RB-associated KRAB zinc finger protein; RGD1562875; Zinc finger protein 769; ZNF769 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
57786 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NYW8 | |
RBAK | |
Synthetic peptide directed towards the middle region of human RBAK. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction