Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBBP4 Mouse anti-Human, Clone: 11G10, Abnova™

Mouse monoclonal antibody raised against human RBBP4.

Manufacturer:  Abnova Corporation MAB12526

Catalog No. 89-934-845


Add to cart

Description

Description

This gene encodes a ubiquitously expressed nuclear protein which belongs to a highly conserved subfamily of WD-repeat proteins. It is present in protein complexes involved in histone acetylation and chromatin assembly. It is part of the Mi-2 complex which has been implicated in chromatin remodeling and transcriptional repression associated with histone deacetylation. This encoded protein is also part of co-repressor complexes, which is an integral component of transcriptional silencing. It is found among several cellular proteins that bind directly to retinoblastoma protein to regulate cell proliferation. This protein also seems to be involved in transcriptional repression of E2F-responsive genes. Three transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Specifications

RBBP4
11G10
Mouse monoclonal antibody raised against human RBBP4.
In PBS.
NURF55/RBAP48
Mouse
IgG2b
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Unconjugated
Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user.
Liquid
RBBP4
Recombinant protein corresponding to amino acids 1-425 of human RBBP4.
Protein G purification
RUO
Primary
5928
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit