Learn More
RBBP4/RbAp48 Mouse anti-Human, DyLight™ 550, Clone: 13D10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP141201R
Catalog No. NBP141201R
Add to cart
Description
Description
RBBP4/RbAp48 Monoclonal antibody specifically detects RBBP4/RbAp48 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
RBBP4/RbAp48
13D10
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
CAF-1 subunit C, CAF-I p48, Chromatin assembly factor 1 subunit C, Chromatin assembly factor I p48 subunit, chromatin assembly factor/CAF-1 p48 subunit, histone-binding protein RBBP4, MSI1 protein homolog, Nucleosome-remodeling factor subunit RBAP48, NURF55, RbAp48, RBBP-4, retinoblastoma binding protein 4, Retinoblastoma-binding protein 4CAF-I 48 kDa subunit, Retinoblastoma-binding protein p48
Mouse
IgG2b
Store at 4°C in the dark.
Monoclonal
This antibody will recognize human RbAp48.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
DyLight™ 550
50 mM sodium borate with 0.05% sodium azide
RBBP4
The complete coding region (amino acids 1-425) of RbAp48 expressed in E. coli.
0.1 mL
Primary
5928
Human