Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBBP4/RbAp48 Mouse anti-Human, DyLight™ 650, Clone: 13D10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP141201C

Catalog No. NBP141201C


Add to cart

Description

Description

RBBP4/RbAp48 Monoclonal antibody specifically detects RBBP4/RbAp48 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

RBBP4/RbAp48
13D10
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Conjugated, Purified
RBBP4
The complete coding region (amino acids 1-425) of RbAp48 expressed in E. coli.
Protein G purified
Cancer, Cell Cycle and Replication, Chromatin Research, Growth and Development, Stem Cell Markers, Tumor Suppressors
Primary
5928.0
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
DyLight™ 650
50 mM sodium borate with 0.05% sodium azide
CAF-1 subunit C, CAF-I p48, Chromatin assembly factor 1 subunit C, Chromatin assembly factor I p48 subunit, chromatin assembly factor/CAF-1 p48 subunit, histone-binding protein RBBP4, MSI1 protein homolog, Nucleosome-remodeling factor subunit RBAP48, NURF55, RbAp48, RBBP-4, retinoblastoma binding protein 4, Retinoblastoma-binding protein 4CAF-I 48 kDa subunit, Retinoblastoma-binding protein p48
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
This antibody will recognize human RbAp48.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit