Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBBP6 Rabbit anti-Human, DyLight™ 350, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP149542UV

Catalog No. NBP149542UV


Add to cart

Description

Description

RBBP6 Polyclonal antibody specifically detects RBBP6 in Human samples. It is validated for Western Blot.
Specifications

Specifications

RBBP6
DyLight™ 350
50 mM sodium borate with 0.05% sodium azide
E3 ubiquitin-protein ligase RBBP6, EC 6.3.2.-, MY038, P2PR, P2P-R, p53-associated cellular protein of testis, PACTDKFZp761B2423, Proliferation potential-related protein, Protein P2P-R, RB-binding Q-protein 1, RBQ1, RBQ-1, retinoblastoma binding protein 6, Retinoblastoma-binding protein 6DKFZp686P0638, Retinoblastoma-binding Q protein 1, SNAMA
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human
Western Blot
Western Blot
Affinity Purified, Conjugated
RBBP6
A synthetic peptide made to an internal portion of the human RBBP6 protein (between residues 20-70) [UniProt Q7Z6E9]
Immunogen affinity purified
Tumor Suppressors
Primary
5930.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit