Learn More
RBBP9 Mouse anti-Human, Clone: OTI2G2, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA501676
Catalog No. 50-167-2800
Add to cart
Description
Description
RBBP9 may play a role in the transformation process due to its capacity to confer resistance to the growth-inhibitory effects of TGF-beta1 through interaction with retinoblastoma and the subsequent displacement of E2F-1.The protein encoded by this gene is a retinoblastoma binding protein that may play a role in the regulation of cell proliferation and differentiation. Two alternatively spliced transcript variants of this gene with identical predicted protein products have been reported, one of which is a nonsense-mediated decay candidate.
Specifications
Specifications
RBBP9
Monoclonal
Unconjugated
RBBP9
Liquid
RBBP9
Full length human recombit protein of human RBBP9 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2G2
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O75884
BOG, RBBP10
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10741