Learn More
Grainger RBBR Tread on Steel Core Wheel 10
Supplier:  Grainger 16V335
Encompass_Preferred
The Flat Free Solid Rubber Wheel by Grainger is a 10 inch diameter by 2.5 inch wide solid rubber wheel with a 1 inch bore size and 1/2 inch tread depth. It has a load capacity of 350 lbs and is made of high quality rubber that is cut, tear and puncture resistant. This solid rubber wheel requires no maintenance or air pressure checks, making it ideal for carts, dollies and other material handling applications. The rubber compound provides good traction and a smooth ride over uneven surfaces. Overall an excellent quality solid rubber wheel for industrial use.
Catalog No. 50-276-6426
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.