Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBFA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598665
Description
RBFA Polyclonal Antibody for Western Blot, IHC (P), ELISA
RBFA is a protein coding gene and also known as Ribosome Binding Factor A.
Specifications
RBFA | |
Polyclonal | |
Unconjugated | |
RBFA | |
1110032A13Rik; AI595940; C18orf22; HsT169; Putative ribosome-binding factor A, mitochondrial; Rbfa; RGD1311910; ribosome binding factor A; ribosome binding factor A (putative) | |
Rabbit | |
Protein G | |
RUO | |
307235, 68731, 79863 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q6P3B9, Q8N0V3 | |
RBFA | |
Recombinant Human Putative ribosome-binding factor A, mitochondrial protein (201-343AA). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction