Learn More
Aviva Systems Biology Corp RBJ Antibody - middle region (ARP56938_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56938_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBJ. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RBJ. Alternate names: DKFZp434N211, RabJS, RBJApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-62039