Learn More
Aviva Systems Biology Corp RBL1 Antibody - N-terminal region (P100682_P050) | 100uL
Supplier:  Aviva Systems Biology Corp P100682_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RBL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RBL1. Alternate names: CP107, MGC40006, PRB1, p107ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62681