Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBM12 Monoclonal Antibody (1D12)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA520938
Description
Peptide Sequence: GPGPIHIGGP PGFASSSGKP GPTVIKVQNM PFTVSIDEIL DFFYGYQVIP GSVCLKYNEK GMPTGEAMVA FESRDEATAA VIDLNDRPIG SRKVKLVLG.
This gene encodes a protein that contains several RNA-binding motifs, potential transmembrane domains, and proline-rich regions. This gene and the gene for copine I overlap at map location 20q11.21. Alternative splicing in the 5' UTR results in two transcript variants. Both variants encode the same protein.
Specifications
RBM12 | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q9NTZ6 | |
RBM12 | |
RBM12 (NP_006038.2, 834 a.a. ~ 932 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
1D12 | |
Unconjugated | |
RBM12 | |
5730420G12Rik; 9430070C08Rik; AI852903; Copine I; Copine-1; Cpne1; HRIHFB2091; KIAA0765; mKIAA0765; RBM12; RNA binding motif protein 12; RNA-binding motif protein 12; RNA-binding protein 12; SH3/WW domain anchor protein in the nucleus; SWAN | |
Mouse | |
Affinity chromatography | |
RUO | |
10137 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction