Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBM14 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548232
Description
Predicted to react with mouse, rat, rabbit, pig, chicken and bovine based on sequence homology.
Isoform 1 may function as a nuclear receptor coactivator, enhancing transcription through other coactivators such as NCOA6 and CITED1. Isoform 2, functions as a transcriptional repressor, modulating transcriptional activities of coactivators including isoform 1, NCOA6 and CITED1.Specifications
RBM14 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q96PK6 | |
RBM14 | |
KLH conjugated synthetic peptide between 563-594 amino acids from the C-terminal region of human RBM14. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
RBM14 | |
RNA-binding protein 14, Paraspeckle protein 2, PSP2, RNA-binding motif protein 14, RRM-containing coactivator activator/modulator, Synaptotagmin-interacting protein, SYT-interacting protein, RBM14, SIP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10432 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction