Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM14 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP180243

 View more versions of this product

Catalog No. NBP180243


Add to cart

Description

Description

RBM14 Polyclonal antibody specifically detects RBM14 in Human, Mouse, Rat, Porcine, Bovine, Canine, Guinea Pig, Rabbit samples. It is validated for Western Blot.
Specifications

Specifications

RBM14
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
RBM14
Synthetic peptide directed towards the N terminal of mouse RBM14. Peptide sequence KEADAKAAIAQLNGKEVKGKRINVELSTKGQKKGPALAIQSGDKTKKPGA.
63 kDa
100ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Guinea pig: 86%.
Bovine, Canine, Guinea Pig, Human, Mouse, Porcine, Rabbit, Rat
Western Blot
Western Blot 1:1000
NP_063922
COAAMGC31756, Paraspeckle protein 2, PSP2DKFZp779J0927, RNA binding motif protein 14, RNA-binding motif protein 14, RNA-binding protein 14, RRM-containing coactivator activator/modulator, SIPMGC15912, Synaptotagmin-interacting protein, SYT-interacting protein, SYTIP1, TMEM137, transmembrane protein 137
Rabbit
IgG
Protein A purified
RUO
Primary
10432
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit