Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ RBM18 Human Recombinant Protein
Highly purified. Generating reliable and reproducible results.
$344.00 - $976.00
Specifications
Concentration | 1mg/mL |
---|---|
For Use With (Application) | SDS-Page |
Formulation | Phosphate buffered saline (pH7.4) containing 10% glycerol |
Gene ID (Entrez) | 92400 |
Molecular Weight | TMW: 24kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25186701
|
Novus Biologicals™
NBP2518670.1MG |
0.1mg |
Each for $344.00
|
|
NBP25186705
|
Novus Biologicals™
NBP2518670.5MG |
0.5mg |
Each for $976.00
|
|
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-190 of Human RBM18 The Recombinant Human RBM18 Protein is derived from E. coli. The Recombinant Human RBM18 Protein has been validated for the following applications: SDS-Page.Specifications
1mg/mL | |
Phosphate buffered saline (pH7.4) containing 10% glycerol | |
TMW: 24kDa | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
RBM18 | |
Yes |
SDS-Page | |
92400 | |
>85%, by SDS-PAGE | |
MGC2734, RNA binding motif protein 18, RNA-binding motif protein 18 | |
Unlabeled |
For research use only and is not approved for use in humans or in clinical diagnosis
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title