Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Rbm20 Polyclonal Antibody

Rabbit Polyclonal Antibody
$249.00 - $454.65
Specifications
| Antigen | Rbm20 |
|---|---|
| Concentration | 0.5 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Western Blot |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119402
![]() |
Invitrogen™
PA5119402 |
200 μL |
$566.00
$454.65 / Each Save $111.35 20% Off |
|
|||||
|
PIPA5121650
|
Invitrogen™
PA5121650 |
20 μL |
Each for $249.00
|
|
|||||
Description
Rbm20 Polyclonal Antibody for Western Blot
This gene encodes a protein that likely binds RNA. Mutations in this gene have been associated with familial dilated cardiomyopathy.Specifications
| Rbm20 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Pig | |
| Rbm20 | |
| 1110018J23Rik; 2010003H22Rik; AI646761; probable RNA-binding protein 20; Rbm20; RNA binding motif protein 20; RNA-binding motif protein 20; RNA-binding protein 20 | |
| Rbm20 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q5T481 | |
| 100157563, 282996 | |
| Recombinant protein RNA Binding Motif Protein 20. The antigen corresponds to amino acid range 1089-1227 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title