Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBM3 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | RBM3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP182420UL
|
Novus Biologicals
NBP18241120UL |
20 μL |
Each for $152.22
|
|
NBP182411
|
Novus Biologicals
NBP182411 |
100 μL |
Each for $436.00
|
|
Description
RBM3 Polyclonal specifically detects RBM3 in Mouse samples. It is validated for Western Blot.Specifications
RBM3 | |
Polyclonal | |
Rabbit | |
NP_058089 | |
5935 | |
Synthetic peptide towards Rbm3. Peptide sequence LFVGGLNFNTDEQALEDHFSSFGPISEVVVVKDRETQRSRGFGFITFTNP. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
IS1-RNPL, putative RNA-binding protein 3, RNA binding motif (RNP1, RRM) protein 3, RNA binding motif protein 3, RNA-binding motif protein 3, RNPL | |
RBM3 | |
IgG | |
Affinity Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title