Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM41 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549029

Catalog No. PA549029

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May bind RNA.
Specifications

Specifications

RBM41
Polyclonal
PBS with 0.09% sodium azide
Q96IZ5, Q8JZV4
RNA-binding protein 41, RNA-binding motif protein 41, RBM41
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
237073, 55285
Western Blot
Unconjugated
RBM41
Liquid
RBM41
KLH conjugated synthetic peptide between 316-343 amino acids from the C-terminal region of human RBM41.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit