Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBM47 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541889
Description
Peptide sequence: HFTSREDAVH AMNNLNGTEL EGSCLEVTLA KPVDKEQYSR YQKAARGGGA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
The RBM47 gene encodes RNA-binding protein 47, which plays an important role in RNA processing and regulation. This protein is involved in various aspects of RNA metabolism, including RNA splicing, editing, stabilization, and translation. RBM47 influences gene expression by binding to specific RNA sequences, affecting post-transcriptional modifications that are critical for cellular development and function. It has been implicated in cancer biology, where changes in RBM47 expression or function can contribute to tumor suppression and progression. RBM47 is known for its role in maintaining the fidelity of RNA editing processes, impacting how cells respond to genetic or environmental changes. Research into RBM47 focuses on its involvement in RNA-mediated regulatory pathways and its potential as a therapeutic target in diseases where RNA processing errors occur, such as certain cancers and genetic disorders.
Specifications
RBM47 | |
Polyclonal | |
Unconjugated | |
RBM47 | |
9530077J19Rik; BC61; hypothetical protein; hypothetical RNA binding protein RGD1359713; NET18; RBM47; RGD1359713; RNA binding motif protein 47; RNA-binding motif protein 47; RNA-binding protein 47; si:ch211-241e15.2; wu:fj94a03 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54502 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
A0AV96 | |
RBM47 | |
Synthetic peptide directed towards the middle region of human RBM47. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction