Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBMY1D Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572104
Description
RBMY1D Polyclonal Antibody for Western Blot
RNA-binding protein which may be involved in spermatogenesis. Required for sperm development, possibly by participating in pre-mRNA splicing in the testis.
Specifications
RBMY1D | |
Polyclonal | |
Unconjugated | |
RBMY1D | |
hRBMY; RBM1; RBM2; RBMY1A1; RBMY1D; RNA binding motif protein, Y-linked, family 1, member D; RNA-binding motif protein 1; RNA-binding motif protein 2; RNA-binding motif protein, Y chromosome, family 1 member A1; RNA-binding motif protein, Y chromosome, family 1 member D; Y chromosome RNA recognition motif 1; YRRM1; YRRM2 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
378949 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P0C7P1 | |
RBMY1D | |
KLH conjugated synthetic peptide between 444-470 amino acids from the C-terminal region of human RBMY1D. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction