Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBP1 Mouse anti-Human, Clone: OTI2H3, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502755S

Catalog No. 50-167-3775


Add to cart

Description

Description

This gene encodes the carrier protein involved in the transport of retinol (vitamin A alcohol) from the liver storage site to peripheral tissue. Vitamin A is a fat-soluble vitamin necessary for growth, reproduction, differentiation of epithelial tissues, and vision. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

RBP1
Monoclonal
1.2 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P09455
CRABP-I, CRBP, CRBP1, CRBPI, RBPC
Mouse
IgG2a
30 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5947
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2H3
Unconjugated
RBP1
Liquid
RBP1
Full length human recombit protein of human RBP1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit