Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBP1 Mouse anti-Human, Clone: OTI2E1, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525533

Catalog No. PIMA525533


Add to cart

Description

Description

The cellular retinol binding proteins (CRBPs) are an important component of retinoid activity, and involved in the transport and metabolism of retinoids. In humans, three known isoforms exist: CRBP1, CRBP2 and CRBP3. Although the function of the retinol binding proteins are not fully understood, it is thought they assist in the formation of retinyl esters for storage or conversion of retinol to retinoic acid. Cellular retinol-binding protein-1 (CRBP-1, RBP1 or Retinol Binding Protein 1) is involved in the transport of Vitamin A from the liver to the peripheral tissue. CRBP-1 is highly expressed in liver cells.
Specifications

Specifications

RBP1
Monoclonal
0.94 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P09455
CRABP-I, CRBP, CRBP1, CRBPI, RBPC, CRBP-I, cellular retinol-binding protein I, retinol-binding protein 1, retinol-binding protein 1, cellular
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5947
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2E1
Unconjugated
RBP1
Liquid
RBP1
Full length human recombit protein of RBP1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit