Sino Biological™ RBP4 Recombinant Human Protein Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human RBP4, including signal sequence (Met 1-Leu 201), fused to polyhistidine tag at C-terminus

Manufacturer: Sino Biological™ 10354H08H250

 View more versions of this product

Catalog No. 10-354-H08H25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Common Name Retinol binding protein 4, plasma
    Molecular Weight 23kd
    Purity ≥85% by SDS-PAGE
    Quantity 5 x 50μg
    Source Human
    Structural Form Recombinant
    Endotoxin Concentration <1 EU/μg
    Gene Symbol RBP7
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Other proteins
    Expression System Mammalian
    Product Type RBP4 Recombinant Human Protein
    For Use With (Application) Analysis of protein structure and performing functional assays
    Format Lyophilized
    Gene ID (Entrez) 5950

    Expressed from DNA sequence encoding human RBP4, including signal sequence (Met 1-Leu 201), fused to polyhistidine tag at C-terminus

    • Measured by its ability to bind all-trans retinoic acid
    • Binding of retinoic acid results in quenching of tryptophan fluorescence in RBP4
    • Recombinant human RBP4 can bind >1.0µM all-trans retinoic acid
    • Lyophilized in PB

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit