Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBP4/Retinol-Binding Protein 4 Antibody (423619), mFluor Violet 610 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB34761MFV610
Description
RBP4/Retinol-Binding Protein 4 Monoclonal antibody specifically detects RBP4/Retinol-Binding Protein 4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
| RBP4/Retinol-Binding Protein 4 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 423619 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, retinol-binding protein 4, plasma | |
| Mouse myeloma cell line NS0-derived recombinant mouse RBP4/Retinol-Binding Protein 4, Met1-Leu201, Accession # NP_035385 | |
| 0.1 mL | |
| Cancer, Diabetes Research, Lipid and Metabolism | |
| 5950 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction