Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ RBP4/Retinol-Binding Protein 4 Antibody Pair [HRP]

Mouse Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2795835PLATES

 View more versions of this product

Catalog No. NB010206


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human RBP4/Retinol-Binding Protein 4.
Specifications

Specifications

Sandwich ELISA
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human RBP4/Retinol-Binding Protein 4.
23.44 pg/mL
5950
RUO
RBP4/Retinol-Binding Protein 4
Cancer, Diabetes Research, Lipid and Metabolism
Human
RBP4
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1500 pg/mL is recommended.
HRP
Antibody Pairs
Sandwich ELISA
  • Detection Antibody: 0.2 mg/mL of mouse anti-Human RBP4/Retinol-Binding Protein 4 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 1 mg/mL of mouse anti-Human RBP4/Retinol-Binding Protein 4 monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 25 ng of recombinant Human RBP4/Retinol-Binding Protein 4.
  • Storage is content dependent.
    Mouse
    5 Plates
    23.44-1500 pg/mL
    interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, retinol-binding protein 4, plasma
    Conjugated
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit