Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBP4/Retinol-BindingProtein4 Mouse anti-Human, Janelia Fluor 549, Clone: 2B4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP126373JF549

Catalog No. NBP12373549

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

RBP4/Retinol-BindingProtein4Monoclonalantibody specifically detects RBP4/Retinol-BindingProtein4 in Human samples.
Specifications

Specifications

RBP4/Retinol-BindingProtein4
2B4
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
RBP4
Recombinant human RBP4 (19-201aa) purified from E. coli
Protein G purified
Cancer, Diabetes Research, Lipid and Metabolism
Primary
5950
Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, retinol-binding protein 4, plasma
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit