Learn More
Abnova™ RBP5 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00083758T01
Catalog No. 89-959-614
Add to cart
Description
Description
Specifications
Specifications
293T
NM_031491.1
pCMV-RBP5 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
RBP5 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 14.96kDa
Transient overexpression cell lysate was tested with anti-RBP5 antibody (Mfr. No. H00083758-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human