Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBPJ Rat anti-Human, Clone: 7A11, Active Motif™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

Specifications

Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Antigen RBPJ
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3113 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
61074
10 μg N/A
50-199-3112 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
61073
100 μg N/A
Description

Description

CSL plays a role in the regulatory network through which muscle cells coordinate their structural and functional states during growth, adaptation, and repair. The protein is abundantly expressed in heart and skeletal muscle and expression is increased by passive stretch. Mutations in this gene can lead to X-linked deafness.
Specifications

Specifications

Monoclonal
Primary
RBPJ
Unconjugated
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit