Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBPMS, Guinea Pig anti-Human,Mouse, Polyclonal, Novus Biologicals
Guinea Pig Polyclonal Antibody
Manufacturer: Novus Biologicals NBP280389
Description
RBPMS Polyclonal antibody specifically detects RBPMS in Human, Mouse Samples. It is validated for Western Blot, ELISA.Specifications
RBPMS | |
1 mg/mL | |
Western Blot 1:500-1:1000, ELISA 1:1000-1:5000 | |
Affinity Purified | |
RBPMS | |
Synthetic peptide within human RBPMS 1-30 conjugated to Keyhole Limpet Haemocyanin. | |
Peptide affinity purified | |
RUO | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Polyclonal | |
Human, Mouse |
ELISA, Western Blot | |
Unconjugated | |
1X TBS(pH7.4), 1%BSA, 50%Glycerol | |
FLJ32971, Heart and RRM expressed sequence, Hermes, HERMESRNA-binding protein with multiple splicing, RBP-MS, RNA binding protein with multiple splicing | |
Guinea Pig | |
IgG | |
100 uL | |
DNA Repair, Mismatch Repair | |
Primary | |
11030 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title