Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBX1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | RBX1 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
RBX1 Polyclonal specifically detects RBX1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
RBX1 | |
Polyclonal | |
Rabbit | |
Cancer, Cell Cycle and Replication, Hypoxia, Neuroscience, Wnt Signaling Pathway, Zinc Finger | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
9978 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:MAAAMDVDTPSGTNSGAGKKRFEVKKWNAVALWAWDIVVDNCAICRNHIMDLCIECQANQASATSEECTVAWGVCNHAFHFHCISRWLKTRQVCPLDNREWEF | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
BA554C12.1, EC 6.3.2.-, FLJ60363, MGC13357, Protein ZYP, Rbx1, Regulator of cullins 1, RING box protein 1, RING finger protein 75, ring-box 1, ring-box 1, E3 ubiquitin protein ligase, RING-box protein 1, RNF75MGC1481, ROC1E3 ubiquitin-protein ligase RBX1, ZYP protein | |
RBX1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title