Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RCCD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559294
Description
Immunogen sequence: PFIAVQPFPA LLDLPMGSDA VKASCGSRHT AVVTRTGELY TWGWGKYGQL GHEDTTSLDR PRRVEYFVDK QLQVKAVTCG PWNTYVYAVE KG Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.
Plays a role in transcriptional repression of satellite repeats, possibly by regulating H3K36 methylation levels in centromeric regions together with KDM8 (PubMed:24981860). Possibly together with KDM8, is involved in proper mitotic spindle organization and chromosome segregation (PubMed:24981860). Plays a role in regulating alpha-tubulin deacetylation and cytoskeletal microtubule stability, thereby promoting cell migration and TGF-beta-induced epithelial to mesenchymal transition (EMT), potentially through the inhibition of KDM8 (PubMed:28455245).
Specifications
RCCD1 | |
Polyclonal | |
Unconjugated | |
RCCD1 | |
RCC1 domain containing 1; RCC1 domain-containing protein 1; RCCD1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91433 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NED2 | |
RCCD1 | |
Recombinant protein corresponding to Human RCCD1. Recombinant protein control fragment (Product #RP-98162). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction