Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ RCGD 423
Click to view available options
Quantity:
50 mg
Description
RCGD 423 is a gp130 modulator. Binds gp130, promotes Myc expression, increases lactate production and activates downstream JAK/STAT signaling pathways in hair follicle stem cells. Also increases articular chondrocyte activation and facilitates joint repair by regulating cartilage growth and differentiation.
Specifications
Specifications
Chemical Name or Material | N-(4-Bromophenyl)-4-phenyl-2-thiazolamine |
CAS | 108237-91-8 |
Quantity | 50 mg |
Molecular Formula | C15H11BrN2S |
Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction