Learn More
RCI Conductivity Indicators
Ideal for comparing various solutions across a wide range of conductivity levels.
$50.15 - $72.00
Specifications
Display Type | LED |
---|---|
For Use With (Application) | Ideal for comparing various solutions across a wide range of conductivity levels |
Sensor Types | Conductivity |
Voltage | 9V |
Requires: 9V transistor battery
Description
Simply immerse the probes in the solution and record the display lamp lit. The higher the numbered display lamp lit, the greater the solution conductivity.
- Unique, multiple lamp conductivity display determines the relative conductivity of solutions
- Choose from the 10-level RCI Junior or the advanced, 20-level RCI-DX conductivity indicator
Specifications
LED | |
Ideal for comparing various solutions across a wide range of conductivity levels | |
Conductivity | |
9V |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.